Assay Details
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609259.3 | XP_005609316.2 | 345 | 91 | ||
XM_023639368.1 | XP_023495136.1 | 1029 | 91 | |||
XM_023639369.1 | XP_023495137.1 | 939 | 91 | |||
XM_023639370.1 | XP_023495138.1 | 3162 | 91 | |||
XM_023639371.1 | XP_023495139.1 | 9532 | 91 | |||
XM_023639372.1 | XP_023495140.1 | 748 | 91 | |||
XM_023639373.1 | XP_023495141.1 | 435 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023639368.1, XM_023639369.1, XM_023639370.1, XM_023639371.1, XM_023639372.1, XM_023639373.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
