Assay Details
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496039.6 | XP_001496089.1 | 1327 | 71 | ||
XM_014738110.2 | XP_014593596.1 | 1268 | 71 | |||
XM_023632757.1 | XP_023488525.1 | 1248 | 71 | |||
XM_023632758.1 | XP_023488526.1 | 1236 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
