Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599967.3 | XP_005600024.3 | 417 | 72 | ||
XM_023619060.1 | XP_023474828.1 | 419 | 72 | |||
XM_023619061.1 | XP_023474829.1 | 419 | 72 | |||
XM_023619062.1 | XP_023474830.1 | 422 | 72 | |||
XM_023619063.1 | XP_023474831.1 | 400 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
