Assay Details
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621435.1 | XP_023477203.1 | 960 | 68 | ||
XM_023621436.1 | XP_023477204.1 | 677 | 68 | |||
XM_023621437.1 | XP_023477205.1 | 588 | 68 | |||
GenBank mRNA | JL616086.1 | - | 6 - 7 | 736 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
