Assay Details
Gene Symbol: ATP5F1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005612872.3 | XP_005612929.1 | 1374 | 63 | ||
GenBank mRNA | HQ890188.1 | - | 3 - 4 | 457 | 63 | |
JL637603.1 | - | 6 - 7 | 940 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
