Assay Details
Gene Symbol: PBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628884.1 | XP_023484652.1 | 927 | 104 | ||
XM_023628885.1 | XP_023484653.1 | 925 | 104 | |||
XM_023628886.1 | XP_023484654.1 | 2787 | 104 | |||
GenBank mRNA | JL618790.1 | - | 3 - 4 | 472 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
