Assay Details
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130486.2 | NP_001123958.1 | 2 - 3 | 349 | 114 | |
GenBank mRNA | CD466015.1 | - | 2 - 3 | 349 | 114 | |
DN507624.1 | - | 2 - 3 | 326 | 115 | ||
EF397508.1 | - | 2 - 3 | 183 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
