Assay Details
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492266.6 | XP_001492316.1 | 519 | 82 | ||
GenBank mRNA | JL620912.1 | - | 2 - 3 | 172 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
