Assay Details
Gene Symbol: ZNF148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601921.3 | XP_005601978.1 | 1083 | 77 | ||
XM_023623598.1 | XP_023479366.1 | 1112 | 77 | |||
XM_023623599.1 | XP_023479367.1 | 1508 | 77 | |||
XM_023623600.1 | XP_023479368.1 | 2644 | 77 | |||
XM_023623601.1 | XP_023479369.1 | 1053 | 77 | |||
XM_023623602.1 | XP_023479370.1 | 1031 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
