Assay Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600435.3 | XP_005600492.1 | 366 | 122 | ||
XM_023620081.1 | XP_023475849.1 | 591 | 122 | |||
XM_023620082.1 | XP_023475850.1 | 704 | 122 | |||
XM_023620083.1 | XP_023475851.1 | 357 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
