Assay Details
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735282.2 | XP_014590768.1 | 1616 | 69 | ||
XM_023626746.1 | XP_023482514.1 | 249 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014735282.2, XM_023626746.1 transcripts.
Set Membership: |
![]() ![]() |
