Assay Details
Gene Symbol: UCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498417.4 | XP_001498467.2 | 596 | 54 | ||
XM_005612045.3 | XP_005612102.1 | 369 | 54 | |||
XM_014741874.2 | XP_014597360.1 | 621 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
