Assay Details
Gene Symbol: PPM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627837.1 | XP_023483605.1 | 1120 | 81 | ||
XM_023627838.1 | XP_023483606.1 | 1160 | 81 | |||
GenBank mRNA | JL620109.1 | - | 1 - 2 | 111 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
