Assay Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503765.6 | XP_001503815.1 | 193 | 68 | ||
XM_005609887.2 | XP_005609944.1 | 198 | 68 | |||
XM_023640844.1 | XP_023496612.1 | 202 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
