Assay Details
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501152.6 | XP_001501202.1 | 525 | 59 | ||
XM_005614302.3 | XP_005614359.1 | 370 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
