Assay Details
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267603.3 | NP_001254532.1 | 3 - 4 | 268 | 80 | |
GenBank mRNA | JQ713943.1 | - | 3 - 4 | 271 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
