Assay Details
Gene Symbol: PRKAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163983.2 | NP_001157455.2 | 7 - 8 | 429 | 88 | |
XM_005611072.3 | XP_005611129.2 | 608 | 88 | |||
GenBank mRNA | DQ318140.1 | - | 6 - 7 | 404 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
