Assay Details
Gene Symbol: MXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638507.1 | XP_023494275.1 | 591 | 93 | ||
XM_023638508.1 | XP_023494276.1 | 477 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
