Assay Details
Gene Symbol: MRPL47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601823.3 | XP_005601880.1 | 595 | 69 | ||
XM_014732764.2 | XP_014588250.1 | 393 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
