Assay Details
Gene Symbol: KLF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363309.4 | XP_003363357.3 | 1270 | 63 | ||
GenBank mRNA | JL622830.1 | - | 1 - 2 | 89 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
