Assay Details
Gene Symbol: IL4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081774.2 | NP_001075243.1 | 9 - 10 | 993 | 95 | |
XM_005598731.3 | XP_005598788.2 | 1476 | 95 | |||
XM_005598732.3 | XP_005598789.2 | 1240 | 95 | |||
XM_005598733.3 | XP_005598790.2 | 1054 | 95 | |||
XM_005598734.3 | XP_005598791.2 | 1132 | 95 | |||
GenBank mRNA | AY081138.1 | - | 9 - 10 | 995 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
