Assay Details
Gene Symbol: EEF1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081785.1 | NP_001075254.1 | 3 - 4 | 260 | 75 | |
GenBank mRNA | AB292250.1 | - | 3 - 4 | 260 | 75 | |
JL615965.1 | - | 1 - 2 | 82 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
