Assay Details
Gene Symbol: EEF1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081785.1 | NP_001075254.1 | 6 - 7 | 667 | 64 | |
GenBank mRNA | AB292250.1 | - | 6 - 7 | 667 | 64 | |
JL615965.1 | - | 4 - 5 | 489 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
