Assay Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081798.1 | NP_001075267.1 | 3 - 4 | 676 | 125 | |
XM_005606254.3 | XP_005606311.2 | 1063 | 125 | |||
XM_014736498.2 | XP_014591984.2 | 1073 | 125 | |||
XM_023630397.1 | XP_023486165.1 | 1266 | 125 | |||
XM_023630398.1 | XP_023486166.1 | 1120 | 125 | |||
XM_023630399.1 | XP_023486167.1 | 659 | 125 | |||
GenBank mRNA | DQ266434.1 | - | 3 - 4 | 676 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
