Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081819.2 | NP_001075288.2 | 1 - 2 | 223 | 82 | |
GenBank mRNA | AB035735.1 | - | 1 - 2 | 223 | 82 | |
EU438779.1 | - | 1 - 2 | 214 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
