Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081819.2 | NP_001075288.2 | 3 - 4 | 318 | 84 | |
XM_005603490.1 | XP_005603547.1 | 451 | 84 | |||
GenBank mRNA | AB035735.1 | - | 3 - 4 | 318 | 84 | |
EU438779.1 | - | 3 - 4 | 309 | 84 | ||
KX345377.1 | - | 3 - 4 | 280 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
