Assay Details
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081826.3 | NP_001075295.1 | 3 - 4 | 339 | 72 | |
GenBank mRNA | AB001692.1 | - | 3 - 4 | 319 | 72 | |
CX601547.1 | - | 3 - 4 | 277 | 72 | ||
GU808339.1 | - | 3 - 4 | 340 | 72 | ||
U38956.1 | - | 3 - 4 | 240 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
