Assay Details
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081831.1 | NP_001075300.1 | 3 - 4 | 397 | 64 | |
XM_023632814.1 | XP_023488582.1 | 581 | 64 | |||
XM_023632815.1 | XP_023488583.1 | 581 | 64 | |||
XM_023632816.1 | XP_023488584.1 | 581 | 64 | |||
XM_023632817.1 | XP_023488585.1 | 581 | 64 | |||
XM_023632818.1 | XP_023488586.1 | 581 | 64 | |||
XM_023632819.1 | XP_023488587.1 | 367 | 64 | |||
GenBank mRNA | AB177987.1 | - | 3 - 4 | 397 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
