Assay Details
Gene Symbol: TYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081840.2 | NP_001075309.2 | 2 - 3 | 792 | 67 | |
XM_014735365.2 | XP_014590851.1 | 944 | 67 | |||
GenBank mRNA | AF076781.1 | - | 1 - 2 | 164 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
