Assay Details
Gene Symbol: CASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081842.1 | NP_001075311.1 | 2 - 3 | 340 | 56 | |
GenBank mRNA | AF090119.1 | - | 2 - 3 | 340 | 56 | |
JL638736.1 | - | 3 - 4 | 524 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
