Assay Details
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081849.1 | NP_001075318.1 | 1 - 2 | 367 | 130 | |
XM_005596086.3 | XP_005596143.1 | 1237 | 130 | |||
GenBank mRNA | AF175709.1 | - | 1 - 2 | 367 | 130 | |
HM569606.1 | - | 1 - 2 | 366 | 130 | ||
X99438.1 | - | 353 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
