Assay Details
Gene Symbol: NR5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081851.1 | NP_001075320.1 | 6 - 7 | 1296 | 110 | |
GenBank mRNA | AF203911.1 | - | 6 - 7 | 1296 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
