Assay Details
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081854.3 | NP_001075323.1 | 2 - 3 | 190 | 75 | |
NM_001085424.1 | NP_001078893.1 | 2 - 3 | 190 | 75 | ||
XM_014741479.2 | XP_014596965.1 | 936 | 75 | |||
GenBank mRNA | AF249307.1 | - | 2 - 3 | 190 | 75 | |
AF257471.1 | - | 2 - 3 | 190 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
