Assay Details
Gene Symbol: CCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081863.2 | NP_001075332.1 | 2 - 3 | 232 | 73 | |
GenBank mRNA | AF506970.1 | - | 2 - 3 | 242 | 73 | |
DN503876.1 | - | 2 - 3 | 262 | 73 | ||
DN506500.1 | - | 2 - 3 | 157 | 73 | ||
EU744564.1 | - | 2 - 3 | 187 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
