Assay Details
Gene Symbol: RPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081885.2 | NP_001075354.1 | 3 - 4 | 321 | 92 | |
XM_005610257.3 | XP_005610314.1 | 471 | 92 | |||
XM_005610258.3 | XP_005610315.1 | 471 | 92 | |||
GenBank mRNA | AY113682.1 | - | 3 - 4 | 323 | 92 | |
JL616351.1 | - | 4 - 5 | 367 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
