Assay Details
Gene Symbol: RPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081885.2 | NP_001075354.1 | 5 - 6 | 701 | 102 | |
XM_005610257.3 | XP_005610314.1 | 851 | 102 | |||
GenBank mRNA | AY113682.1 | - | 5 - 6 | 703 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
