Assay Details
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081896.1 | NP_001075365.1 | 3 - 4 | 339 | 59 | |
XM_014734200.1 | XP_014589686.1 | 392 | 59 | |||
GenBank mRNA | AY373339.1 | - | 3 - 4 | 339 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
