Assay Details
Gene Symbol: NFYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081900.2 | NP_001075369.2 | 5 - 6 | 579 | 74 | |
XM_023630974.1 | XP_023486742.1 | 747 | 74 | |||
XM_023630975.1 | XP_023486743.1 | 769 | 74 | |||
GenBank mRNA | AY499345.1 | - | 5 - 6 | 579 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
