Assay Details
Gene Symbol: TPMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081905.1 | NP_001075374.1 | 5 - 6 | 495 | 65 | |
XM_023624007.1 | XP_023479775.1 | 723 | 65 | |||
XM_023624008.1 | XP_023479776.1 | 727 | 65 | |||
GenBank mRNA | AY827076.1 | - | 5 - 6 | 495 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
