Assay Details
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081906.1 | NP_001075375.1 | 4 - 5 | 708 | 103 | |
GenBank mRNA | AY919871.1 | - | 4 - 5 | 708 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
