Assay Details
Gene Symbol: MBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081916.1 | NP_001075385.1 | 2 - 3 | 298 | 75 | |
XM_014732958.1 | XP_014588444.1 | 498 | 75 | |||
GenBank mRNA | DQ372924.1 | - | 2 - 3 | 298 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
