Assay Details
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081928.2 | NP_001075397.1 | 2 - 3 | 144 | 113 | |
GenBank mRNA | AF257470.1 | - | 2 - 3 | 158 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
