Assay Details
Gene Symbol: PMP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081936.1 | NP_001075405.1 | 2 - 3 | 207 | 62 | |
XM_005597020.2 | XP_005597077.1 | 494 | 62 | |||
XM_005597021.3 | XP_005597078.1 | 380 | 62 | |||
GenBank mRNA | AY280708.1 | - | 2 - 3 | 207 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
