Assay Details
Gene Symbol: AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081937.1 | NP_001075406.1 | 121 | 54 | ||
GenBank mRNA | AY339891.1 | - | 121 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
