Assay Details
Gene Symbol: HSP90AB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081938.1 | NP_001075407.1 | 6 - 7 | 1060 | 66 | |
XM_023624011.1 | XP_023479779.1 | 976 | 66 | |||
GenBank mRNA | AB043676.1 | - | 5 - 6 | 932 | 66 | |
AY383484.1 | - | 6 - 7 | 1060 | 66 | ||
HQ890170.1 | - | 1 - 2 | 179 | 66 | ||
JL624672.1 | - | 1 - 2 | 306 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
