Assay Details
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081939.1 | NP_001075408.1 | 2 - 3 | 205 | 85 | |
GenBank mRNA | AY499344.1 | - | 2 - 3 | 205 | 85 | |
HQ890206.1 | - | 2 - 3 | 243 | 85 | ||
JL641793.1 | - | 2 - 3 | 262 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
