Assay Details
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081946.2 | NP_001075415.2 | 4 - 5 | 522 | 55 | |
GenBank mRNA | M69020.1 | - | 4 - 5 | 519 | 55 | |
U21127.1 | - | 4 - 5 | 523 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
