Assay Details
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081947.1 | NP_001075416.1 | 2 - 3 | 203 | 113 | |
XM_005603498.3 | XP_005603555.1 | 606 | 113 | |||
XM_005603499.3 | XP_005603556.1 | 196 | 113 | |||
XM_005603500.3 | XP_005603557.1 | 479 | 113 | |||
GenBank mRNA | JL616060.2 | - | 1 - 2 | 107 | 113 | |
M91161.1 | - | 2 - 3 | 203 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
