Assay Details
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081947.1 | NP_001075416.1 | 4 - 5 | 459 | 80 | |
XM_005603498.3 | XP_005603555.1 | 862 | 80 | |||
XM_005603499.3 | XP_005603556.1 | 452 | 80 | |||
XM_005603500.3 | XP_005603557.1 | 735 | 80 | |||
GenBank mRNA | JL616060.2 | - | 3 - 4 | 363 | 80 | |
M91161.1 | - | 4 - 5 | 459 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
