Assay Details
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081947.1 | NP_001075416.1 | 5 - 6 | 599 | 130 | |
XM_005603498.3 | XP_005603555.1 | 130 | ||||
XM_005603499.3 | XP_005603556.1 | 130 | ||||
XM_005603500.3 | XP_005603557.1 | 130 | ||||
GenBank mRNA | HQ889952.1 | - | 1 - 2 | 130 | ||
JL616060.2 | - | 4 - 5 | 130 | |||
M91161.1 | - | 5 - 6 | 599 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
